missing translation for 'onlineSavingsMsg'
Learn More

Itaconic anhydride, 97%

Product Code. 11470718
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. Quantity unitSize
11470718 500 g 500g
11450718 25 g 25g
11460718 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11470718

Brand: Thermo Scientific Alfa Aesar B25216.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2170-03-8 | C5H4O3 | 112.08 g/mol

Itaconic anhydride is used in immobilization of β-glucosidase. It undergoes estrification to obtain suitable monomers for emulsion polymerization itaconates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Itaconic anhydride is used in immobilization of β-glucosidase. It undergoes estrification to obtain suitable monomers for emulsion polymerization itaconates.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2170-03-8
Molecular Formula C5H4O3
Molecular Weight (g/mol) 112.08
MDL Number MFCD00005530
InChI Key OFNISBHGPNMTMS-UHFFFAOYSA-N
Synonym itaconic anhydride, itaconic acid anhydride, 2-methylenesuccinic anhydride, methylenesuccinic anhydride, 2,5-furandione, dihydro-3-methylene, itaconicanhydride, succinic anhydride, methylene, dihydro-3-methylene-2,5-furandione, 3-methylenedihydro-2,5-furandione, unii-y455ks1u7q
PubChem CID 75110
IUPAC Name 3-methylideneoxolane-2,5-dione
SMILES C=C1CC(=O)OC1=O

Specifications

Melting Point 68°C to 72°C
Boiling Point 114°C to 115°C (12 mmHg)
Quantity 500 g
Beilstein 2212
Solubility Information Hydrolyzes in water.
Formula Weight 112.09
Percent Purity 97%
Chemical Name or Material Itaconic anhydride
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.