missing translation for 'onlineSavingsMsg'
Learn More

Itaconic anhydride, 98%

Product Code. 10433551 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10433551 25 g Glass bottle 25g
10228433 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10433551 Supplier Thermo Scientific Acros Supplier No. 412940250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2170-03-8 | C5H4O3 | 112.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2170-03-8
Molecular Formula C5H4O3
Molecular Weight (g/mol) 112.08
MDL Number MFCD00005530
InChI Key OFNISBHGPNMTMS-UHFFFAOYSA-N
Synonym itaconic anhydride, itaconic acid anhydride, 2-methylenesuccinic anhydride, methylenesuccinic anhydride, 2,5-furandione, dihydro-3-methylene, itaconicanhydride, succinic anhydride, methylene, dihydro-3-methylene-2,5-furandione, 3-methylenedihydro-2,5-furandione, unii-y455ks1u7q
PubChem CID 75110
IUPAC Name 3-methylideneoxolane-2,5-dione
SMILES C=C1CC(=O)OC1=O

Specifications

Melting Point 66.0°C to 70.0°C
Color White to Cream
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 98%
Beilstein 17, 442
Formula Weight 112.08
Percent Purity 98%
Chemical Name or Material Itaconic anhydride
Show More Show Less
compliance-icons
Product Identifier
  • Itaconic anhydride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.