missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Itraconazole
GREENER_CHOICE

Product Code. 18745836
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18745836 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18745836 Supplier Tocris Bioscience™ Supplier No. 5981/50
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

SMO antagonist; acts at different binding site to cyclopamine (Cat No. 1623)

Itraconazole is a SMO antagonist (IC50 = 690 nM); acts at different binding site to cyclopamine (Cat. No. 1623). Also cytochrome p450 inhibitor (IC50 = 16-26 nM). Inhibits cell cycle at G1 phase in vitro and blocks angiogenesis in vivo (IC50 = 160 nM). Antifungal.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[4-[4-[4-[[2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-1-piperazinyl]phenyl]-2,4-dihydro-2-(1-methylpropyl)-3H-1,2,4-triazol-3-one
CAS 84625-61-6
Quantity 50 mg
Target Smoothened Receptor Antagonists
Molecular Formula C35H38Cl2N8O4
Formula Weight 705.63
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.