missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ivabradine hydrochloride
GREENER_CHOICE

Product Code. 18767596
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18767596 10 mg 10mg
18706326 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18767596 Supplier Tocris Bioscience™ Supplier No. 6542/10
  • ONLINE EXCLUSIVE PRICE
    86.00 €
    80.85 € / 10mg
    Save 5.15 €
    5% Off
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

HCN channel blocker; inhibits If pacemaker current

Ivabradine hydrochloride is a HCN channel blocker (IC50 is approximately 0.5 - 2.5 μM). Bradycardic agent that inhibits If pacemaker current in sinoatrial node cells.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[3-[[[(7S)-3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl]methylamino]propyl]-1,3,4,5-tetrahydro-7,8-dimethoxy-2H-3-benzazepin-2-one hydrochloride
CAS 148849-67-6
Quantity 10 mg
Molecular Formula C27H36N2O5.HCl
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.