missing translation for 'onlineSavingsMsg'
Learn More

Ivabradine N-oxide (Mixture of Diastereomers), TRC

Product Code. 30395885
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30395885 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395885 Supplier Toronto Research Chemicals Supplier No. TRCI94052025MG
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ivabradine N-Oxide
CAS 2511244-97-4
Molecular Formula C27 H36 N2 O6
Quantity 25 mg
Synonym 3-(7,8-Dimethoxy-2-oxo-1,2,4,5-tetrahydro-3H-benzo[d]azepin-3-yl)-N-(((S)-3,4-dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl)methyl)-N-methylpropan-1-amine Oxide, Ivabradine N-Oxide, 2H-3-Benzazepin-2-one, 3-[3-[[[(7S)-3,4-dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl]methyloxidoamino]propyl]-1,3,4,5-tetrahydro-7,8-dimethoxy- (ACI), 3-[3-[[[(7S)-3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl]methylox
InChI Formula InChI=1S/C27H36N2O6/c1-29(31,17-21-11-20-14-25(34-4)26(35-5)16-22(20)21)10-6-8-28-9-7-18-12-23(32-2)24(33-3)13-19(18)15-27(28)30/h12-14,16,21H,6-11,15,17H2,1-5H3/t21-,29?/m1/s1
SMILES COc1cc2CCN(CCC[N+](C)([O-])C[C@H]3Cc4cc(OC)c(OC)cc34)C(=O)Cc2cc1OC
IUPAC Name N-[[(7S)-3,4-dimethoxy-7-bicyclo[4.2.0]octa-1,3,5-trienyl]methyl]-3-(7,8-dimethoxy-4-oxo-2,5-dihydro-1H-3-benzazepin-3-yl)-N-methylpropan-1-amine oxide
Molecular Weight (g/mol) 484.584
Formula Weight 484.257
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.