missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ IWP L6

Description
IWP L6 is a potent inhibitor of Porcupine (PORCN), a membrane-bound O-acyltransferase (MBOAT) (EC50 = 0.5 nM). Shown to inhibit Wnt-dependent processes such as tailfin regeneration and posterior axis formation in zebrafish; also reduces branching morphogenesis in cultured mouse embryonic kidneys.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | IWP L6 |
| Purity | >98% |
| Molecular Weight (g/mol) | 472.58 |
| Inhibitors | IWP L6 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?