missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (±)-J 113397
GREENER_CHOICE

Product Code. 30001416
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001416 50 mg 50mg
18074952 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001416 Supplier Tocris Bioscience™ Supplier No. 2598/50
  • ONLINE EXCLUSIVE PRICE
    1675.00 €
    1581.30 € / 50mg
    Save 93.70 €
    5% Off
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and selective NOP antagonist

Potent and selective NOP receptor antagonist (IC50 values are 2.3, 1400, 2200 and > 10000 nM for NOP, κ, μ and δ-opioid receptors respectively). Inhibits nociceptin/orphanin FQ-induced hyperalgesia in the mouse tail-flick test.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 217461-40-0
Molecular Formula C24H37N3O2

Specifications

Quantity 50 mg
Formula Weight 399.57
Purity >98%
Molecular Weight (g/mol) 399.579
Chemical Name or Material (±)-J 113397
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.