Learn More
GPE Scientific™ J Young Schlenk Flask, Cylindrical with Fine Thread Tap and Cone
Description
- Solidly constructed for use with vacuum manifolds
- Designed for use with Schlenk lines
- Includes fine thread tap and cone
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| For Use With (Equipment) | Vacuum manifolds and Schlenk lines |
| Includes | Fine thread tap and cone |
| Product Type | Schlenk Flask |
| Socket Joint | Size: 19 |