Learn More
GPE Scientific™ J Young Thumb Screw Lock Clips for Spherical Joints
Click to view available options
For Use With (Equipment):
12/2 and 12/5 size spherical joints
18/7 and 18/9 size spherical joints
28/12 and 28/15 size spherical joints
35/20 size spherical joints
40/25 size spherical joints
Unit Size:
Each
Description
- Thumb screw design
- Securely fasten J Young spherical joints
Specifications
Specifications
| Type | Thumb Screw Lock Clips for J Young Spherical Joints |
| For Use With (Equipment) | 18/7 and 18/9 size spherical joints |