Learn More
GPE Scientific™ J Young Thumb Screw Lock Clips for Spherical Joints
Product Code. 15864904
Change view
Click to view available options
For Use With (Equipment):
12/2 and 12/5 size spherical joints
18/7 and 18/9 size spherical joints
28/12 and 28/15 size spherical joints
35/20 size spherical joints
40/25 size spherical joints
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15864904 40/25 size spherical joints Each
15824904 12/2 and 12/5 size spherical joints Each
15834904 18/7 and 18/9 size spherical joints Each
15844904 28/12 and 28/15 size spherical joints Each
15854904 35/20 size spherical joints Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15864904 Supplier GPE Scientific™ Supplier No. TJC/41
Estimated Shipment: 25-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
GPE Scientific™ J Young Thumb Screw Lock Clips for Spherical Joints lock in place to securely fasten J Young spherical joints.
  • Thumb screw design
  • Securely fasten J Young spherical joints
Specifications
Type Thumb Screw Lock Clips for J Young Spherical Joints
For Use With (Equipment) 40/25 size spherical joints