missing translation for 'onlineSavingsMsg'
Learn More

Brady™ J20 Replacement Cutter Blade

Product Code. 16110501 Shop All Brady Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16110501 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16110501 Supplier Brady™ Supplier No. 150160
Estimated Shipment: 13-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Brady™ J20 Replacement Cutter Blade is a replacement blade that ensures labels keep a clean edge.

  • For use with the BradyJet™ J2000 Color Label Printer

Specifications

Product Type Cutter Tool
For Use With (Equipment) Brady Label Printers
Dimensions (L x W x H) 8 x 0.25 x 4 in.
Format Cutter
Languages Universal
Quantity 1 Ea.
Alternate Part Number J20-CUTTER
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.