missing translation for 'onlineSavingsMsg'
Learn More

Hughes Jacketed and Insulated Tank Shower

Product Code. 17772348
Change view
Click to view available options
Capacity (Metric):
350 L
1500 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17772348 350 L Each
17762348 1500 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17772348 Supplier Hughes Supplier No. HSHJ14K3SS
  • ONLINE EXCLUSIVE PRICE
    10192.85 €
    5662.56 € / Pack of 1
    Save 4530.29 €
    44% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Insulated emergency tank shower with eye/face wash unit, for use in non-hazardous hot climates. Designed to meet and exceed ANSI standards by providing tepid water, even when there is no mains water supply.

  • Insulated emergency tank shower with eye/face wash unit, for use in non-hazardous hot climates
  • Jacketed and insulated models maintain a steady temperature
  • Suitable for use where insufficient water pressure exists
  • Side mounted eye/face wash unit
  • Designed to meet and exceed ANSI standards by providing tepid water for over 15 minutes at a guaranteed flow of 76 liters per minute, even when there is no mains water supply

Specifications

Capacity (Metric) 350 L
Material Stainless Steel
Product Type Tank Shower
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.