missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals JAK2 Inhibitor IV

Product Code. 15496939
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15496939 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15496939 Supplier Thermo Scientific Chemicals Supplier No. J65506.#0
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

 

An aminoindazole compound that potently inhibits the activity of both the wild-type JAK2 and the active V617F mutant

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Description Pale yellow
Content And Storage 2°C to 8°C
For Use With (Application) An aminoindazole compound that potently inhibits the activity of both the wild-type JAK2 and the active V617F mutant
CAS 1110502-30-1
Molecular Formula C17H20N4O2S
Formula Weight 344.40
Synonym 4-(3-Amino-1H-indazol-4-yl)-N-(tert-butyl)benzenesulfonamide;
Sensitivity Light Sensitive
Product Type JAK2 Inhibitor IV
Form Solid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.