missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Janelia Fluor 646, Maleimide

Product Code. 30306106
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30306106 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 30306106 Supplier Tocris Bioscience™ Supplier No. 6590/1
  • ONLINE EXCLUSIVE PRICE
    361.00 €
    341.25 € / 1mg
    Save 19.75 €
    5% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Red dye (Reactive group: maleimide). Application: flow cytometry, confocal microscopy, super resolution microscopy including dSTORM & STED. Suitable for live cell imaging

Key information: Janelia Fluor™ 646, Maleimide is a red fluorogenic fluorescent dye, supplied with a maleimide reactive group for conjugation (thiol reactivity). Suitable for live cell imaging. Application: Suitable for flow cytometry, confocal microscopy, super resolution microscopy (SRM) including dSTORM (in both live and fixed cells) and STED. Can be multiplexed for two color imaging with Janelia Fluor™ 549 SE (Cat. No. 6147). Cell permeable. Properties and Photophysical Data: Excitation and emission maxima (λ) are 646 nm and 664 nm, respectively; quantum yield = 0.54; extinction coefficient = 152,000 M-1cm-1 (measured in ethanol plus 0.1% TFA); A280 correction factor = 0.19; lactone-zwitterion equilibrium constant (KL-Z) = 0.0012. Please see the product protocol for further information and a guide to protein/antibody labeling. To measure the absorbance spectrum of this dye we recommend the following solvent: ethanol or trifluoroethanol plus 0.1% TFA. We also offer Janelia Fluor™ conjugated antibodies and custom conjugation services with our sister company Novus Biologicals. Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(3-(Azetidin-1-ium-1-ylidene)-7-(azetidin-1-yl)-5,5-dimethyl-3,5-dihydrodibenzo[b,e]silin-10-yl)-4-((2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)carbamoyl)benzoate
Quantity 1 mg
Target Sulfhydyl-Reactive Janelia Fluor Dyes
Molecular Formula C35H34N4O5Si
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.