missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Janelia Fluor 646, SE
Descripción
Key Information: Janelia Fluor™ 646, SE is a red fluorogenic fluorescent dye; supplied with an NHS ester reactive group for the labeling of primary amines. Suitable for live cell imaging.Application: Suitable for flow cytometry, confocal microscopy, super resolution microscopy (SRM) techniques including dSTORM (in both live and fixed cells) and STED. Cell permeable.Properties and Photophysical Data: NHS ester can be converted to relevant substrate for use in self-labeling tag systems, e.g. HaloTag™ and SNAP-tag™. Can be multiplexed for two color imaging with Janelia Fluor™ 549 SE (Cat. No. 6147). Excitation and emission maxima (λ) are 646 nm and 664 nm, respectively; quantum yield = 0.54; extinction coefficient = 152,000 M-1cm-1 (measured in TFE plus 0.1% TFA); A280 correction factor = 0.19; lactone-zwitterion equilibrium constant (KL-Z) = 0.0012. Please see the protocol for further information on protein/antibody labeling and conjugation. We recommend that stock solutions of this dye are prepared in anhydrous DMF. To measure the absorbance spectrum of this dye we recommend the following solvent: ethanol or trifluoroethanol plus 0.1% TFA.We also offer Janelia Fluor™ conjugated antibodies and custom conjugation services with our sister company R&D Systems. HaloTag is a trademark of Promega Corporation, and SNAP-tag is a trademark of New England BioLabs, Inc.
Especificaciones
Especificaciones
| Chemical Name or Material | 1-[7-(1-Azetidinyl)-10-[2-carboxy-5-[[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl]phenyl]-9,9-dimethyl-9-silaanthracen-2(9H)-ylidene]azetidinium, inner salt |
| CAS | 1811539-59-9 |
| Quantity | 1 mg |
| Product Line | Tocris |
| Target | Amine-Reactive Janelia Fluor Dyes |
| Molecular Formula | C33H31N3O6Si |
| Purity | 95% |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?