missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Janelia Fluor™ 669, Maleimide

Product Code. 30226370
Change view
Click to view available options
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
30226370 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 30226370 Supplier Tocris Bioscience™ Supplier No. 8097/200U
Stock Up and Save This Summer
Use promo code 27950
View all promotions
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Red dye (Reactive group: maleimide). Application: flow cytometry, confocal microscopy, super resolution microscopy including dSTORM & STED. Suitable for live cell imaging

Key information: Janelia Fluor™ 669, Maleimide is a red fluorogenic fluorescent dye, supplied with a maleimide reactive group for conjugation (thiol reactivity). Suitable for live cell imaging.

Application: Suitable for flow cytometry, confocal microscopy, super resolution microscopy (SRM) including dSTORM (in both live and fixed cells) and STED. Can be multiplexed for two color imaging with Janelia Fluor™ 549 SE (Cat. No. 6147). Cell permeable.

Properties and Photophysical Data: Excitation and emission maxima (λ) are 669 nm and 689 nm, respectively.

Please see the product protocol for further information and a guide to protein/antibody labeling.

To measure the absorbance spectrum of this dye we recommend the following solvent: ethanol or trifluoroethanol plus 0.1% TFA.

We also offer Janelia Fluor™ conjugated antibodies and custom conjugation services with our sister company Novus Biologicals.

Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(3-(Azetidin-1-ium-1-ylidene)-7-(azetidin-1-yl)-5,5-dimethyl-3,5-dihydrodibenzo[b,e]silin-10-yl)-4-((2-((2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)amino)-2-oxoethyl)thio)-3,5,6-trifluorobenzoate
Quantity 200 μg
Target Fluorescent Dyes
Molecular Formula C36H33F3N4O5SSi
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.