missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Janelia Fluor 669, SE

Product Code. 18740538
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18740538 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18740538 Supplier R&D Systems™ Supplier No. 6420/2
  • ONLINE EXCLUSIVE PRICE
    418.45 €
    374.85 € / 2mg
    Save 43.60 €
    10% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Far red dye (Reactive group: NHS ester). Applications: super resolution microscopy e.g. dSTORM & STED, confocal and live cell imaging

Janelia Fluor® 669, SE is a red fluorescent dye; supplied as an NHS ester for coupling to primary amine groups. NHS ester can be converted to relevant substrate for use in self-labeling tag systems, e.g. HaloTag™ and SNAP-tag™. Suitable for confocal fluorescent imaging and super resolution microscopy (SRM) techniques, such as dSTORM (live and fixed cells). Cell permeable. Excitation maximum = 669 nm; emission maximum = 682 nm; Quantum yield = 0.37; Extinction coefficient = 116,000 M-1cm-1; Correction factor = 0.0430. Retains 97% fluorescence after 30 bleaching cycles.

We also offer Janelia Fluor™ conjugated antibodies and custom conjugation services with our sister company R&D Systems.

HaloTag is a trademark of Promega Corporation, and SNAP-tag is a trademark of New England BioLabs, Inc.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[7-(1-Azetidinyl)-10-[2-Carboxy-5-[(2-(2,5-dioxopyrrolidin-1-yl)oxy)-2-oxoethyl]thio-3,4,6-trifluorophenyl]-9,9-dimethyl-9-silaanthracen-2(9 H)-ylidene]azetidinium, inner salt
CAS 2127150-20-1
Quantity 2 mg
Product Line Tocris
Target Amine-Reactive Janelia Fluor™ Dyes
Molecular Formula C34H30F3N3O6SSi
Formula Weight Observed MW: 693.77
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.