missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint Gobain Life Sciences Chemware™ PFA Jar with Cap
Click to view available options
Capacity (Metric):
120 mL
360 mL
480 mL
1000 mL
2000 mL
Unit Size:
Each
Description
- Chemware™ PFA jars store solutions, paste and bulk materials with no concern about leaching, corrosion, contamination or evaporation.
- They are resistant to corrosive liquids, acids, solvents, sensitive reagents, and staining solutions.
- PFA jars feature a large mouth for easy access and non-stick surface for easy cleaning.
- There is an area for identification marking on side of the jars.
- Stackable, these jars have a knurled top for secure grip to facilitate easy opening and closing.
Specifications
Specifications
| Capacity (Metric) | 360 mL |
| Diameter (English) | 3.3 in. |
| For Use With (Application) | Storing Digestion Samples |
| Includes | Cap |
| Material | PFA |
| Weight (Metric) | 0.8 kg |
| Closure Included | Yes |
| Mouth | Wide |
| Shape | Round |
| Height (Metric) | 90.6 mm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction