Learn More
Cole-Parmer™ Jenway™ 100 Disposable Cuvettes
Product Code. 11400559
Change view
Click to view available options
Capacity (Metric):
1.5 to 3.0 mL
2.4 to 4.5 mL
Description:
UV and visible wavelengths
Visible wavelengths only
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Description unitSize
11400559 1.5 to 3.0 mL Visible wavelengths only Pack of 100
11490549 2.4 to 4.5 mL Visible wavelengths only Pack of 100
11420559 2.4 to 4.5 mL UV and visible wavelengths Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11400559 Supplier Cole-Parmer™ Supplier No. 9961034
Only null left
Add to basket
Add to basket
Specifications
Type Cuvette
Description Visible wavelengths only
Capacity (Metric) 1.5 to 3.0 mL
Material Plastic
Quantity 100/Pk.
Path Length 10 mm
For Use With (Application) Spectral Measurement of Liquids