missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Jenway™ 500 Disposable Cuvettes

Product Code. 11410559
Change view
Click to view available options
Capacity (Metric):
2.4 to 4.5 mL
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11410559 2.4 to 4.5 mL Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11410559 Supplier Cole-Parmer™ Supplier No. 8307030
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Type Cuvette
Disposable Single-use
Capacity (Metric) 2.4 to 4.5 mL
Material Plastic
Includes 5 boxes of (060 084) 2.4 to 4.5mL cuvettes for visible wavelengths
Quantity 500/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.