missing translation for 'onlineSavingsMsg'
Learn More

Bibby Scientific™ Jenway™ ATC/Temperature Probes

Product Code. 11430519
Change view
Click to view available options
For Use With (Equipment):
Jenway pH Meters 3150 and 3205
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11430519 Jenway pH Meters 3150 and 3205 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11430519 Supplier Bibby Scientific™ Supplier No. 5953070
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Choose Jenway™ ATC/Temperature Probes to use with Jenway™ Performance pH electrodes, the electrodes of choice for advanced applications where faster response times and greater reproducibility are required.

For use with Jenway™ Performance pH electrodes:
  • Quickly reach equilibrium
  • Significantly reduced response times
  • Far greater reproducibility
  • Significantly reduced long-term drift

Specifications

Connector Types 3150/3205 waterproof connector
Type Sealed Combination with Bulb End
For Use With (Application) Standard measurements of general solutions, field measurements or in education
Includes integrated temperature probe
Length (Metric) 120 mm
Material Polypropylene
pH Range 0 to 14
Temperature (Metric) Range 0°C to 100°C
For Use With (Equipment) Jenway pH Meters 3150 and 3205
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.