Learn More
Cole-Parmer™ Jenway™ Combination Ion Selective Electrode
Product Code. 11480699
Change view
Click to view available options
Filling Solution Type:
Chloride
Nitrate
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Filling Solution Type Unit Size
11480699 Nitrate Each
11430669 Chloride Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11480699 Supplier Cole-Parmer™ Supplier No. 1009015
  • ONLINE EXCLUSIVE PRICE
    1005.00 €
    859.04 € / Pack of 1
    Save 145.96 €
    14% Off
Estimated Shipment: 31-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Jenway™ Combination Ion Selective Electrodes provide efficient and accurate concentration measurement of specific ions.
  • Available for use with chloride or nitrate filling solutions
Specifications
Filling Solution Type Nitrate
Type Combination Ion Selective Electrode
Parameter Nitrate