Learn More
Cole-Parmer™ Jenway™ Dissolved Oxygen Probes
Product Code. 13177614
Change view
Click to view available options
Includes:
Automatic Temperature Control and DIN Connector
Automatic Temperature Control and Waterproof Connector
Mini-DIN Connector
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
13177614 Automatic Temperature Control and DIN Connector Each
13114334 Automatic Temperature Control and Waterproof Connector Each
12801621 Mini-DIN Connector Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13177614 Supplier Cole-Parmer™ Supplier No. 9996471
  • ONLINE EXCLUSIVE PRICE
    635.00 €
    545.13 € / Pack of 1
    Save 89.87 €
    14% Off
Estimated Shipment: 05-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Connections DIN
Includes Automatic Temperature Control and DIN Connector
For Use With (Equipment) Jenways 9500 Dissolved Oxygen Meter