Learn More
Jenway™ pH Electrodes
Use these electrodes for advanced applications where faster response times and greater reproducibility are required.
Brand:  Jenway™ 68006-40
Code : NEW
Additional Details : Weight : 0.08100kg
 View more versions of this product
Product Code. 11449489
  • 94.70 € / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Quickly reach equilibrium
  • Significantly reduced response times
  • Far greater reproducibility
  • Significantly reduced long-term drift
Specifications
Specifications
S7
Sealed Combination with Bulb End and Protective Cover
standard measurements of general solutions, field measurements or in education
Epoxy
0°C to 60°C
12 mm
12 dia. x 120 mm L
120 mm
0 to 14