missing translation for 'onlineSavingsMsg'
Learn More

Bibby Scientific™ Jenway™ Spear Knife Probe

Product Code. 11410629
Change view
Click to view available options
pH Range:
0 to 14
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range unitSize
11410629 0 to 14 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11410629 Supplier Bibby Scientific™ Supplier No. 6800638
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Bibby Scientific™ Jenway™ Spear Knife Probe is epoxy based for the measurement of solid samples.

  • Spear shape allows for pH measurement of more solid sample materials
  • Suitable for use in the food industry for measurement into frozen or semi-frozen foodstuffs, protects electrode from damage

Specifications

Connector Types BNC
Diameter (Metric) 12.7 to 20.0 mm
Type Knife Tipped
For Use With (Application) Food industry for measurement into frozen or semi-frozen foodstuffs, protects electrode from damage
Length (Metric) 95 mm
Material Epoxy body
pH Range 0 to 14
Temperature (Metric) Range 0°C to 80°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.