Learn More
Chemsplash Jet Spray 88 OverBoot with Anti-Slip Sole
Product Code. 30058179
Change view
Click to view available options
Size:
Medium/Large
Unit Size:
100 pairs
Product Code. Size Unit Size
30058179 Medium/Large 100 pairs
1 options
Product Code. 30058179 Supplier Chemsplash Supplier No. 2746.700.007
  • ONLINE EXCLUSIVE PRICE
    708.23 €
    656.36 € / 100 pairs
    Save 51.87 €
    7% Off
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Jet Spray Overboot with Taped Seams Type PB(3-B) with Anti-Slip sole is made from Chemsplash 88 gsm Jet Spray fabric.
Chemsplash Jet Spray Overboot with Taped Seams Type PB (3-B)
  • 88 GSM Chemsplash Jet Spray Fabric
  • Anti-Slip Sole and Anti-Static Strips
  • Low Linting
  • Elasticated Top and Ankles
  • Taped Seams
  • PFAS Free
Description: The Chemsplash Jet Spray Overboot with Taped Seams Type PB (3-B) is made from 88 GSM Chemsplash Jet Spray fabric. It is highly resistant to chemical splash and dust, providing a strong protective barrier. The overboot features a PVC anti-slip and anti-static sole, making it ideal for safe and secure movement in environments with potential hazards. The elasticated top and ankles ensure a secure fit, while the taped seams provide added protection. This overboot is low-linting, helping maintain a clean environment. The Chemsplash Accessories range is designed to protect both the wearer and the environment, offering maximum comfort and wearability.
Specifications
Color Yellow
Material Polyethylene Laminate
Size Medium/Large
Waterproof Yes
Product Type Controlled Environments Boots