Learn More
Chemsplash Jet Spray Surgeons Gown -Taped Seams
Product Code. 30058325
Change view
Click to view available options
Color:
Yellow
Unit Size:
Pack of 100
Product Code. Color Unit Size
30058325 Yellow Pack of 100
1 options
Product Code. 30058325 Supplier Chemsplash Supplier No. 6248.700.999
  • ONLINE EXCLUSIVE PRICE
    1230.00 €
    1105.44 € / Pack of 100
    Save 124.56 €
    10% Off
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Jet Gown with Taped Seams Type PB(3-B) is highly resistant against high hazard permeation offering high levels of protection against even the most corrosive of chemicals.
Chemsplash Jet Gown with Taped Seams
  • Material: 88GSM Chemsplash Jet Spray Fabric
  • High Hazard Protection
  • Ankle Length and Tie Back
  • Double Cuffs - Internally Knitted and Elasticated Outside
  • Double Stand Collar
  • Velcro and Adhesive Tape Fastening on Inside Rear Neck
  • Anti-Static and Non-Linting
  • Taped seams
  • PFAS Free
Description: The Chemsplash Jet Gown with Taped Seams, Type PB(3-B), is highly resistant against high hazard permeation, offering high levels of protection against even the most corrosive chemicals. Made from a thick 88gsm heavy weight Polypropylene/Polyethylene multilayer laminated material. Chemsplash accessories are designed to protect the wearer from contaminants in the environment or to protect the environment and production from body-shedding particles or hair from the wearer. This range has been designed to ensure maximum comfort and wearability while maintaining the highest safety standards.
Specifications
Size Universal
Gender Unisex
No. of Pockets 1
Product Type Controlled Environments Gown
Material Composite Fabric
Color Yellow
Closure Type Ties
Disposable Single-use
Sterility Non-sterile
Cuff Style Knit
Latex Free Yes
Sleeve Style Long Sleeve
Autoclavable Autoclavable