missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ JetQuick™ Blood and Cell Culture DNA Midiprep Kit

Product Code. 15657878
Change view
Click to view available options
No. of Reactions:
20 Preps
50 Preps
Unit Size:
Pack of 20
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
15657878 20 Preps Pack of 20
15667878 50 Preps Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15657878 Supplier Invitrogen™ Supplier No. A30703

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The JetQuick™ Blood and Cell Culture DNA Midiprep Kit is designed for rapid and efficient purification of genomic DNA (gDNA) from blood, body fluids (amniotic fluid, saliva, sperm, lymph), and mammalian cells.

Utilizes silica membrane-based technology that allows for isolation of 20-50 kb DNA that is suitable for a wide range of downstream applications, including PCR, restriction enzyme digestion, and Southern blotting.

Key benefits include:
• Large volume processing, up to 3 mL of blood or saliva
• Long double-stranded DNA (20-50 kb) recovery
• Compatible with centrifugation or a vacuum manifold

The midiprep format enables the processing of up to 3 mL of whole blood or other body fluid. DNA from up to 1 x 108 mammalian cells can be isolated. The purified DNA is ready for downstream analysis.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1 x 66 mL Buffer K1 x Store at Room Temperature
• 1 x 72 mL Buffer K2 x Store at Room Temperature
• 1 x 105 mL Buffer KX, Store at Room Temperature
• 1 x 44 mL Elution Buffer, Store at Room Temperature
• 2 x 1.3 mL Rnase A, Store 2°C to 8°C
• 5 x 26 mg Protease (lyophilized powder), Store 2°C to 8°C
• 4 Spin Column Bags (5 columns/bag)
Format Kit
Isolation Technology Spin Column
Scale Midi
Sample Type Blood, Cells
For Use With (Application) Real-Time PCR, PCR
High-throughput Compatibility Not High-throughput Compatible
No. of Reactions 20 Preps
Product Line JetQuick
Product Type Blood and Cell Culture DNA Kit
Quantity 20 Preps
Shipping Condition Room Temperature
Target Genomic DNA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.