missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ JNJ 16259685
GREENER_CHOICE

Product Code. 18074292
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18074292

Brand: Tocris Bioscience™ 2333/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly potent, mGlu1-selective non-competitive antagonist

Sub-nanomolar potent, non-competitive mGlu1 antagonist (Ki = 0.34 nM). Inhibits glutamate-induced Ca2+ response at the human mGlu1 receptor with an IC50 value of 0.55 nM. Selective over mGlu5 (> 400-fold) and displays no activity at mGlu2, mGlu3, mGlu4, mGlu6, AMPA or NMDA receptors (IC50 > 10 μM). Centrally active following systemic administration.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 409345-29-5
Molecular Formula C20H23NO3

Specifications

Quantity 10 mg
Formula Weight 325.41
Purity >98%
Molecular Weight (g/mol) 325.408
Chemical Name or Material JNJ 16259685
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.