missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ JNJ 55511118

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
JNJ 55511118 is a high affinity and selective negative modulator of AMPA receptors containing TARP-γ8 (Ki = 26 nM). Exhibits <50% binding against a panel of 52 receptors, ion channels and transporters (except 5HT2B and melatonin receptors 78% and 57%, respectively). Also displays minimal activity against other TARP-less AMPARs and AMPARs coexpressed with other TARPs or CNIH2. It reduces alcohol self-administration in a mouse models of chronic drinking. Orally bioavailable and brain penetrant. Anticonvulsant.
Specifications
Specifications
| Chemical Name or Material | 5-[2-Chloro-6-(trifluoromethoxy)phenyl]-1,3-dihydro-2H-benzimidazol-2-one |
| CAS | 2036081-86-2 |
| Assay Percent Range | >98% |
| Quantity | 25 mg |
| Molecular Formula | C14H8ClF3N2O2 |
| Formula Weight | Observed MW: 328.67 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction