missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ JNJ 55511118
GREENER_CHOICE

Product Code. p-7114682
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18719817

Brand: Tocris Bioscience™ 6278/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective, high affinity negative modulator of AMPA receptors containing TARP-γ8

JNJ 55511118 is a high affinity and selective negative modulator of AMPA receptors containing TARP-γ8 (Ki = 26 nM). Exhibits <50% binding against a panel of 52 receptors, ion channels and transporters (except 5HT2B and melatonin receptors 78% and 57%, respectively). Also displays minimal activity against other TARP-less AMPARs and AMPARs coexpressed with other TARPs or CNIH2. It reduces alcohol self-administration in a mouse models of chronic drinking. Orally bioavailable and brain penetrant. Anticonvulsant.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-[2-Chloro-6-(trifluoromethoxy)phenyl]-1,3-dihydro-2H-benzimidazol-2-one
CAS 2036081-86-2
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C14H8ClF3N2O2
Formula Weight Observed MW: 328.67
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.