missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ JP-2-196
Click to view available options
:
50 mg
Description
JP-2-196 is a covalent RNF126 chemical handle that binds to several other RING E3 ubiquitin ligases including RNF14, RNF40, RNF219, MID2, and LRSAM. By facilitating E3 ligase recruitment, JP-2-196 induces proteasomal degradation of diverse target proteins. It serves as a modular scaffold for the design and synthesis of molecular glue degraders.
Specifications
Specifications
| Chemical Name or Material | 1-(4-Methoxyphenyl)-4-(1-piperazinyl)-2-butene-1,4-dione hydrochloride |
| Quantity | 50 mg |
| Target | Molecular Glues |
| Molecular Formula | C15H18N2O3.HCl |
| Purity | 0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction