missing translation for 'onlineSavingsMsg'
Learn More

(+)-JQ-1, MedChemExpress

Product Code. 16488207
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5mg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488207 5 mg 5mg
16456076 100 mg 100mg
16445498 10 mg 10mg
16421656 500 mg 500mg
16421066 200 mg 200mg
16401677 50 mg 50mg
16401329 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 16488207 Supplier Medchem Express Supplier No. HY13030
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (+)-JQ-1 (JQ1) is a potent, specific, and reversible BET bromodomain inhibitor, with IC50s of 77 and 33 nM for the first and second bromodomain (BRD4(1/2)). (+)-JQ-1 also activates autophagy.

(+)-JQ-1 (JQ1) is a potent, specific, and reversible BET bromodomain inhibitor, with IC50s of 77 and 33 nM for the first and second bromodomain (BRD4(1/2)). (+)-JQ-1 also activates autophagy.

Specifications

Chemical Name or Material (+)-JQ-1
CAS 1268524-70-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H25ClN4O2S
Quantity 5 mg
Synonym JQ1
Solubility Information DMSO : ≥ 45 mg/mL (98.47 mM)
SMILES O=C(C[C@H]1C2=NN=C(N2C3=C(C(C4=CC=C(C=C4)Cl)=N1)C(C)=C(S3)C)C)OC(C)(C)C
Molecular Weight (g/mol) 456.99
Formula Weight 456.99
Percent Purity 98.65%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.