missing translation for 'onlineSavingsMsg'
Learn More

JSH-150, MedChemExpress

Product Code. 16482698
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482698 5 mg 5mg
16450489 10 mg 10mg
16424136 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16482698 Supplier Medchem Express Supplier No. HYX0150

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JSH-150 is a highly selective and potent CDK9 inhibitor with an IC50 of 1 nM.

JSH-150 is a highly selective and potent CDK9 inhibitor with an IC50 of 1 nM.

Specifications

Chemical Name or Material JSH-150
CAS 2247481-21-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H33ClN6O2S
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (197.99 mM; Need ultrasonic)
SMILES COCCN[C@@H]1CC[C@@H](NC2=NC=C(Cl)C(C3=CSC(NCC4(C#N)CCOCC4)=N3)=C2)CC1
Molecular Weight (g/mol) 505.08
Formula Weight 505.08
Percent Purity 95.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.