Learn More
Tocris Bioscience™ JTE 607 Dihydrochloride
Active metabolite binds CPSF3; Pro-drug
Brand:  Tocris Bioscience™ 5185/10
Encompass
Product Code. 15716556
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
JTE 607 is a pro-drug that is cleaved by carboxylesterase 1 (CES1) to its active metabolite, which then binds to cleavage and polyadenylation specificity factor 3 (CPSF3; Kd = 370 nM at human CPSF3). Treatment of A-673 cells with JTE 607 results in inhibition of mRNA cleavage and accumulation of nuclear R-loops. JTE 607 induces apoptosis in leukemia cells in vitro and prolongs survival in a mouse leukemia model.
JTE 607 also inhibits cytokine release; it inhibits production of IL-1β, IL-8, IL-6, IL-10 and TNFα (IC50 values are 5.9, 7.3, 8.8, 9.1 and 11.0 nM, respectively) from LPS-stimulated PBMCs. In a rat model of lung injury, JTE 607 reduces proinflammatory cytokine-release and attenuates lung permeability.
Specifications
Specifications
JTE 607 dihydrochloride
N-[3,5-Dichloro-2-hydroxy-4-[2-(4-methyl-1-piperazinyl)ethoxy]benzoyl]-L-phenylalanine ethyl ester dihydrochloride
Soluble to 100mM in water and to 100mM in DMSO
Bioactive Small Molecule
188791-09-5
10 mg
Solid
Product Suggestions
Product Suggestions
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Product Certifications
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Research Use Only. Not For Human or Veterinary Use