missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Juglanin

Product Code. 16491728
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491728 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16491728 Supplier MedChemExpress Supplier No. HYN3442
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Juglanin, a natural occurring flavonoid, is a JNK acticator, with inflammation and anti-tumor activities. Juglanin can induce apoptosis and autophagy on human breast cancer cells.

Specifications

Content And Storage 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Molecular Weight (g/mol) 418.35 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 5041-67-8
Product Type Natural Products
Purity 96.0%
Solubility DMSO : 100 mg/mL (239.03 mM∣Need ultrasonic)
Target JNK∣Apoptosis∣Autophagy
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.