missing translation for 'onlineSavingsMsg'
Learn More

Julolidine hydrobromide, 97%, Thermo Scientific™

Product Code. 15487637
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487637 100 g 100g
15467637 5 g 5g
15477637 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15487637 Supplier Thermo Scientific Alfa Aesar Supplier No. H36298.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83646-41-7
Molecular Formula C12H16BrN
Molecular Weight (g/mol) 254.17
MDL Number MFCD00198009
InChI Key KHWBRFVBPGPEOJ-UHFFFAOYSA-N
Synonym julolidine hydrobromide, 2,3,6,7-tetrahedron-1h,5h-pyrido 3,2,1-ij quinoline hydrobromide, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizine hydrobromide, labotest-bb lt00451605, 2,3,6,7-tetrahydro-1h-5h-benzo i,j quinolizine, hbr, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinoline hydrobromide
PubChem CID 12255670
IUPAC Name 1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene hydrobromide
SMILES Br.C1CN2CCCC3=C2C(C1)=CC=C3

Specifications

Melting Point 239°C to 242°C
Color Pink
Quantity 100 g
UN Number UN2923
Sensitivity Hygroscopic
Formula Weight 254.17
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Julolidine hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.