missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Jumbosep™ Centrifugal Device Membrane Inserts
Description
- Typically concentrates 60 mL sample volumes to 5 mL in 30 minutes
- Achieve 50X concentration and > 90% recovery in just minutes
- Low protein-binding Omega™ membrane and polysulfone housing minimize losses due to non-specific binding
- Versatile Omega membrane is available in a variety of MWCOs
- Color-coded and laser etched for easy identification
- Built-in dead stop prevents spinning to dryness
- Unique sealing mechanism prevents retentate leakage and filtrate contamination
- Economical - Sample reservoir and filtrate receiver can be sanitized or autoclaved, and reused
Specifications
Specifications
| Color | Red |
| Description | EFA: 15.2 cm2 |
| Material (Membrane) | PES |
| MWCO | 30 kDa |
| Sterility | Non-sterile |
| Quantity | 12/Pk. |
| Volume (Metric) | 20 to 60 mL (Sample) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?