Learn More
Cytiva Jumbosep™ Spare Parts and Accessories - Filtrate Receiver and Cap
Description
- Utilizes proprietary high-performance multi-layer filter membranes
- Plates offer time savings strong performance claims and streamlined workflow improvements
- Each plate comes with a V-shaped bottom collection plate and lid
Specifications
Specifications
| Description | EFA: 15.2 cm2 |
| For Use With (Equipment) | Jumbosep Centrifugal Device |
| Sterility | Non-sterile |
| Quantity | 12/Pk. |
| Volume (Metric) | 20 to 60 mL (Sample) |