missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Jump-In™ CHO-K1 Kit

Product Code. 11632089
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11632089 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11632089 Supplier Invitrogen™ Supplier No. A14148
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Jump-In™ CHO-K1 Kit

The Jump-In™ CHO-K1 Kit allows the targeted integration of genetic material into a specific pre-engineered R4 site in the Jump-In™ CHO-K1 cell line, to create an isogenic stable cell line with less effort and in less time than traditional cell engineering methods.

The high retargeting efficiency, made possible by the R4 sites in the CHO-K1 cell line, allows the use of the isogenic pool for additional experiments without the need for clonal selection. Alternatively, the high retargeting efficiency allows for the easy selection of a positive stable clone expressing your gene of interest.

The Jump-In™ CHO-K1 Kit Lets You:

  • Quickly and efficiently develop stably engineered isogenic cell pools in about half the time compared to traditional cell engineering methods
  • Utilize isogenic expression from a defined genomic locus as the ideal solution for comparative analysis of gene families, isoforms, or orthologs
  • Generate multiple cell lines in parallel using the simplified work flow
  • Easily access the technology without complicated licenses or restrictions to interpret

    Save Time with Rapid and Efficient Generation of Engineered Cell Lines
    With the Jump-In™ CHO-K1 Kit you can generate functional cell pools in as little as 2 weeks without laborious clone isolation and analysis, and the streamlined protocol makes it easier to generate several cell lines at the same time. Even generation of clonal cell lines can be done with reduced time and effort due to the high percentage of positive clones. In addition, the Jump-In™ technology gives you the freedom to generate an unlimited number of cell lines without restrictive licensing requirements.

    Expand Your Experimental Capabilities
    The Jump-In™ CHO-K1 Kit is the ideal solution for cells and assays where transient engineering technologies are problematic, as well as for difficult to The kit also provides a convenient way to create target panels of gene families, isoforms, or orthologs. Genes coding for large proteins or multi-unit proteins are not a problem since the Gateway™ destination vectors accept large inserts.

    The kit includes:
  • pJTI™ R4 Dest CMV pA vector (100μg)
  • pJTI™ R4 Int vector (100μg)
  • Jump-In™ CHO-K1 Cells (2 vials @ 1 ml each)

    For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Product Line Jump-In
    Content And Storage The kit includes:
    Jump-In™ CHO-K1 cells
    pJTI™ R4 Int (integrase vector)
    pJTI™ R4 DEST CMV pA (destination vector)

    Upon receipt, immediately store the cells in liquid nitrogen and the vectors at -20 °C. Cells stored at -80°C can quickly lose viability.
    Product Type CHO-K1 Kit
    Quantity 1 kit
    Cell Line Jump-In™ CHO-K1
    Format Kit
    Expression System Mammalian
    Key Functions Stable Cell Line Development, Targeted Integration
    Promoter CMV
    Selection Agent (Eukaryotic) Hygromycin
    System Type Jump-In™
    Show More Show Less
    I am planning to generate a Jump-In platform cell line. Do you recommend mapping the site of integration and checking against the database to pick a clone where integration has occurred in a "good" hot spot?

    We would recommend engineering an expression marker/reporter in the plasmid used to create the platform line, and then screening the platform line for expression of this marker to identify a high-expressing locus. Otherwise, the process can get quite labor-intensive, as multiple lines would have to be screened after retargeting.

    Does the pJTI PhiC31 Int vector contain a nuclear localization signal (NLS), and would adding an NLS increase the efficiency of site-specific integration at pseudo attP sites?

    The pJTI Phic31 Int vector does not contain an NLS. Adding an NLS could increase the efficiency of site-specific integration at pseudo attP sites, but there are no data to support it. There is one paper describing the use of an NLS on a PhiC31 integrase vector, but the authors didn't measure integration into pseudo attP sites.

    In the Jump-In system, how much DNA or what controls do I need to include in order to get one integration event?

    The amount of DNA to be used to obtain single copies should be determined by control experiments done in the absence of integrase. The same amount of DNA that yields less than 5 colonies in the absence of integrase should be used in the presence of integrase. Typically, the integrase expression plasmid makes up most of the amount of DNA used for transfection.

    When should I use the Jump-In Fast system versus the Jump-InTI system?

    We recommend using the Jump-In Fast system if you need stable mammalian expression and want to quickly generate well-expressing clones. You can have well-expressing clones with one or more integrations at the PhiC31 pseudo-att P sites. A Southern blot is necessary to confirm the number of integrated events. Use the Jump-InTI system if you need isogenic expression, where every cloned gene would be expressed from the same locus in the same background, with no chromosomal position effects.

    What is the difference between the Jump-In and Flp-In systems?

    The Jump-In system is PhiC31-integrase mediated and is a stable, targeted, and irreversible mammalian expression system. It consists of the Jump-In Fast system that involves a single integration step and the Jump-InTI (targeted integration) system that needs two integration steps, both of which are targeted and irreversible. In contrast, the Flp-In system is a stable, targeted mammalian expression system that is reversible. The first integration is random (integration of pFRT/lacZeo), and the second integration (integration of the Flp-In expression vector) is targeted but reversible.

    When should I consider reversible integration (Flp-In system) vs irreversible integration (Jump-In system)?

    Use irreversible integration (Jump-In system) if the transgene should be sustained in the mammalian genome for a long time. Use reversible integration such as Flp-In system if the transgene needs to be replaced with another gene of interest after a short period of time.

    What controls do I need in the Jump-In system to check for the successful retargeting of the platform line?

    The second step in targeted integration is the retargeting event mediated by the R4 integrase where the genetic elements of interest are site-specifically transferred from the retargeting expression construct (created using the MultiSite Gateway Pro module) onto the genome of the platform line. This integration event also positions the EF1alpha promoter upstream of the blasticidin, neomycin, or eosin resistance gene (i.e., "promoterless" selection marker), thus allowing the selection of transformants that are successfully "retargeted" using the appropriate selection agent. Although you select from successfully retargeted clones using the blasticidin, Geneticin, or Zeocin antibiotic, you may also perform a nested PCR to amplify the region from the EF1alpha promoter to the appropriate resistance gene. You can amplify the hygromycin resistance gene as a positive control. Similar to the platform line creation, you may also perform a Southern blot analysis with a probe designed for your gene of interest.

    What controls do I need in the Jump-In system to check for the presence of the R4 target site after the creation of the platform cell line?

    A platform cell line is created when the R4 attP retargeting sequences are site-specifically inserted into the mammalian genome via PhiC31 Int-mediated recombination. In addition to the R4 retargeting sequences, this integration event introduces the hygromycin resistance gene under the control of the HSV TK promoter and the promoterless Bsd, Neo, or Zeo resistance marker, depending on the platform vector used (i.e., pJTI/Bsd, pJTI/Neo, or pJTI/Zeo). Although you select for transformants carrying the R4 retargeting sequences by their resistance to hygromycin, you may perform PCR analysis to check the integrity of the R4 attP retargeting sequences. For this, we recommend amplifying the region from the R4 attP sequence to the appropriate resistance marker (depending on the platform line used) using the genomic DNA from the platform line. A nested PCR is recommended to reduce the high background you may observe with only primary PCR. Alternatively, you may create a labeled DNA probe by PCR amplifying an approximately 1.5 kb region covering the retargeting sequences, and then perform a Southern blot analysis. The Southern blot will also act as an additional check to verify that only a single copy of the retargeting sequence is integrated into the genome.

    How much DNA or what controls do I need to include in the Jump-In system in order to get one integration event?

    The amount of DNA to be used to obtain single copies should be determined by control experiments done in the absence of integrase. The same amount of DNA that yields less than 5 colonies in the absence of integrase should be used in the presence of integrase. Typically, the integrase expression plasmid makes up most of the amount of DNA used for transfection.

    When should I use the Jump-In Fast system vs the Jump-In-TI (Targeted Integration) system?

    Use the Jump-In Fast system if you need stable mammalian expression and want to quickly generate well-expressing clones. You can have well-expressing clones with one or more integrations at the PhiC31 pseudo-att P sites. A Southern blot is necessary to confirm the number of integrated events.

    Use the Jump-In TI system if you need isogenic expression, where every cloned gene would be expressed from the same locus in the same background with no chromosomal position effects.

    What is the difference between Jump-In and Flp-In systems?

    The Jump-In system is PhiC31-integrase mediated and is a stable, targeted, and irreversible mammalian expression system, involving one integration step. The Jump-In TI (Targeted Integration) system needs two integration steps, both of which are targeted and irreversible. In contrast, the Flp-In system is a stable, targeted mammalian expression system that is reversible. The first integration is random (integration of pFRT/lacZeo) and the second integration (integration of the Flp-In expression vector) is targeted but reversible.


    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.