missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Jump-In™ Fast Gateway™ Core Kit

Product Code. 10587364
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10587364 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10587364 Supplier Invitrogen™ Supplier No. A10894

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Jump-In™ Fast Gateway™ Core Kit

The Jump-In™ Fast Gateway™ Core Kit is intended for rapid engineering of mammalian cell lines through the stable and irreversible integration of your gene of interest (GOI) at specific genomic loci (pseudo attP sites). Following stable integration, only 5-10 clones need to be analyzed to identify clones with high expression levels.
The Jump-In™ Fast Gateway™ Core Kit contains the promoter-less pJTI™ Fast DEST expression vector and a vector for transient expression of the PhiC31 integrase.
Please note: the pJTI™ Fast DEST expression vector requires the cloning of a promoter upstream of the gene of interest, which is facilitated by any of the MultiSite Gateway™ Pro Kits.
This kit is also a component of the Jump-In™ Fast Gateway™ System.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The Jump-In™ Fast Gateway™ Core Kit is composed of 2 vectors:
pJTI™ Fast DEST Vector (150 ng⁄μl; total 40 μl),
pJTI™ PhiC31 Int Vector (0.5 μg⁄μl; total 20 μl).

Store vectors at -20°C.
Cloning Method Gateway™
Delivery Type Transfection
Cell Type Mammalian
Format Kit
Promoter None (Promoterless)
Vector Jump-In Vectors, pJTI
Expression System Mammalian
For Use With (Application) Transfection
High-throughput Compatibility High-throughput Compatible
Key Functions Stable Cell Line Development, Targeted Integration, Clone your own Promoter
Product Line Jump-In
Product Type Core Kit
Protein Tag Untagged
Quantity 1 Kit
Selection Agent (Eukaryotic) Hygromycin
Selection Marker Eukaryotic HygroR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.