missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Jump-In™TI™ Platform Kit

Product Code. 10555485
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10555485

Brand: Invitrogen™ A10897

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Jump-In™TI™ Platform Kit

The Jump-In™TI™ Platform Kit enables generation of a Jump-In™TI™ cell line. A Jump-In™TI™ cell line is created by co-transfecting one of three vectors containing the R4 attP target site with the transiently expressed vector for the PhiC31 integrase into your mammalian cell line.
Generating a Jump-In™TI™ cell line is the first step in creating a stable, targeted, and irreversible mammalian cell line expressing your gene of interest. For targeted and irreversible integration of your gene of interest into your Jump-In™TI™ cell line, use the Jump-In™TI™ Gateway™ Vector Kit.
This kit is also a component of the Jump-In™TI™ Gateway™ System.
TRUSTED_SUSTAINABILITY

Specifications

Delivery Type Transfection
Description Jump-In™TI™ Platform Kit, 6μg, Gateway™ Cloning Method, 1 Kit, Mammalian Expression, Enables Generation of a Jump-In™TI™ Cell Line, Also a Component of the Jump-In™TI™ Gateway™ System, Platform Kit Enables Generation of a Jump-In™TI™; Cell Line
Promoter None (Promoterless)
Product Type Jump-In™TI™ Platform Kit
Selection Agent (Eukaryotic) Blasticidin, Geneticin™ (G-418), Zeocin™
Stability Stable
Content And Storage -20°C
Protein Tag Untagged
Quantity 1 Kit
Vector Jump-in vectors, pJTI
For Use With (Application) Protein expression, stem cell engineering and reprogramming
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.