missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Excellent performance, small size, low weight (2 kg) and battery-powered mode - ideal whenever mobility counts (in the laboratory, on-site, or outdoors)
- Photomultiplier tube in single photon counting mode
- High-sensitivity model: <50 amol ATP/tube with a dynamic range >6 orders of magnitude
- Spectral range luminescence: 380 - 630 nm
- Compatible with luminescence tubes (12 x 47 mm, 12 x 55 mm, 12 x 75 mm), microcentrifuge tubes (1, 1.5, and 2 mL) and capped vials, 4 mL (14 x 54 mm)
- Equipped with rechargeable batteries for many hours of independent operation; can be connected to a mains supply for automatic recharging
Specifications
Specifications
| Type | Luminometer |
| Display Type | Matrix |
| Detector | Photomultiplier Tube in Single Photon Counting Mode |
| Detection Method | Luminescence |
| Detection Limits | <1 fmol ATP |
| Color | Light Gray |
| For Use With (Application) | Luminescence Assays |
| Voltage | 110/240 V |
| Frequency | 50/60 Hz |
| For Use With (Equipment) | Tubes |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Sekretesspolicy.
Hittar du en möjlighet till förbättring?