missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ JW 74
GREENER_CHOICE

Product Code. 18717189
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18717189 10 mg 10mg
18727668 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18717189 Supplier Tocris Bioscience™ Supplier No. 5653/10
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Wnt pathway inhibitor

JW 74 is an inhibitor of canonical Wnt signaling (IC50 = 790 nM). Targets the β-catenin destruction complex (GSK-3?/AXIN/APC). Increases AXIN2 and decreases active β-catenin levels in colorectal cancer (CRC) cells. Also arrests cell cycle in G1 and inhibits proliferation of CRC cells in vitro. Inhibits tumor growth and formation in mouse models.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[4-(4-Methoxyphenyl)-5-[[[3-(4-methylphenyl)-1,2,4-oxadiazol-5-yl]methyl]thio]-4H-1,2,4-triazol-3-yl]-pyridine
CAS 863405-60-1
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C24H20N6O2S
Formula Weight Observed MW: 456.52
Purity >99%

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.