Learn More
Tocris Bioscience™ JW 74
GREENER_CHOICE
Wnt pathway inhibitor
Brand:  Tocris Bioscience™ 5653/50
Encompass
Product Code. 15747016
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
JW 74 is an inhibitor of canonical Wnt signaling (IC50 = 790 nM). Targets the β-catenin destruction complex (GSK-3?/AXIN/APC). Increases AXIN2 and decreases active β-catenin levels in colorectal cancer (CRC) cells. Also arrests cell cycle in G1 and inhibits proliferation of CRC cells in vitro. Inhibits tumor growth and formation in mouse models.
Specifications
Specifications
4-[4-(4-Methoxyphenyl)-5-[[[3-(4-methylphenyl)-1,2,4-oxadiazol-5-yl]methyl]thio]-4H-1,2,4-triazol-3-yl]-pyridine
863405-60-1
50 mg
Soluble to 100mM in DMSO