Learn More
uvex K-Basic Extra Aramid Fiber Cut/Heat Protection Gloves
Product Code. 17777609 Shop All Uvex Products
Change view
Click to view available options
Size:
3
8
10
12
Unit Size:
5 pairs
Pack of 2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17777609 3 5 pairs
15346952 8 Pack of 2
15316952 10 Pack of 2
15326952 12 Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17777609 Supplier uvex Supplier No. 6017904
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Course-knit aramid fiber gloves are ideal for applications that involve both mechanical and thermal risks. Suitable for contact heat up to 250°C.
  • Course-knit aramid fiber gloves for applications that involve both thermal and mechanical risks
  • A high level of thermal insulation when in direct contact with warm/hot objects
  • Aramid fiber construction provides resistance against cuts
  • Cotton liner on the inside increases user comfort
  • Ideal for light to medium metalworking, glass handling, and automotive industries
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 388:2016 (2 4 4 2 D) and EN 407:2004 (X 2 X X X X)
Coating Material Uncoated
Color Yellow
Latex Free Yes
Material Aramid Fiber
Product Type Heat-resistant Gloves
Size 3
Style Full-hand Mitt
Temperature (Metric) Max 250°C
Liner Material Cotton
Series K-Basic Extra
Yarn Gauge 7