Learn More
uvex K Junior Ear Muffs
Product Code. 17727549 Shop All Uvex Products
Change view
Click to view available options
Color: Blue
Product Code.
17727549 Color: Blue
1 options
Product Code. 17727549 Supplier uvex Supplier No. 2600010
  • ONLINE EXCLUSIVE PRICE
    26.50 €
    24.63 € / Pack of 1
    Save 1.87 €
    7% Off
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Uvex K protective ear muffs offer an impressive combination of high levels of protection combined with great design.
  • Soft surface
  • Easy to mould
  • Comfortable to wear
  • Low weight
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 352-1
Noise Reduction Rating (NRR) 35 dB
Corded No
Product Type Ear Muffs
Disposable Reusable
Metal Detectable Yes
Headband Color Black
Dielectric No
Style Over-the-head
Foldable Yes