missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descrizione
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Two in one: 2 compartments (vertical seperated) allow the storage of flammable liquids and aggressive chemicals in one cabinet
- User-friendly and safe (folding door compartment): door open arrest system (standard), convenient and safe removal or storage of containers
- Always closed (folding door compartment): serial automatic door closing (TSA) releases the arrested door after approx. 60 seconds; acoustic and visual signal before closing
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible) / cylinder lock
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
- Fire resistance 90 minutes
- type tested in accordance with EN 14470-1
- EN 16121/16122 conformity
- GS certified
- CE compliant
Warranty
Five years
Specifica
Specifica
| Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
| Configuration | 3 Shelves, 1 Perforated Insert (380 mm), 1 Bottom Collecting Sump, 6 Pull-out Shelves |
| Depth (Metric) Exterior | 617 mm |
| Depth (Metric) Interior | 524 mm |
| Dimensions (D x W x H) Interior | Left: 524 x 450 x 1742 mm, Top Right: 542 x 491 x 862 mm, Bottom Right: 542 x 491 x 862 mm |
| Dimensions (D x W x H) | 617 x 1197 x 1969 mm |
| Door Style | Wing, Folding, Auto-close |
| For Use With (Application) | Safe and approved storage of aggressive and flammable hazardous substances in working areas |
| Height (Metric) Exterior | 1969 mm |
| Height (Metric) Interior | 1742 mm |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?