missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Combi Safety Storage Cabinets K-UB-90

Product Code. 15914366 Shop All asecos Products
Change view
Click to view available options
Model:
K90.060.110.050.UB.ST
K90.060.140.050.UB.3T
K90.060.140.050.UB.ST
Weight (Metric):
152.50 kg
186.50 kg
196.50 kg
No. of Doors:
1
3
No. of Shelves:
1
2
Pakningsstørrelse:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Model Weight (Metric) No. of Doors No. of Shelves unitSize
15914366 K90.060.110.050.UB.ST 152.50 kg 1 1 Each
15984366 K90.060.140.050.UB.ST 186.50 kg 1 1 Each
15914376 K90.060.140.050.UB.3T 196.50 kg 3 2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy

For indoor use only

Artikelnummer. 15914366 Leverandør asecos™ Leverandørnr. 3035800135362

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

Denne vare kan ikke returneres. Se returpolicy

For indoor use only

Safe and approved storage of aggressive and flammable hazardous substances in working areas

  • Robust construction and longevity: body made of powder-coated sheet steel
  • User-friendly and safe: door and drawer remain open in any position, interior of the cabinet completely visible — access to all containers, 93° opening of the wing door
  • Safe: drawer fitted with earthing wire incl. clamp as standard, seal welded sump, drawers are self-closing in the event of fire
  • No unauthorised use: door and drawer lockable with cylinder lock and locking state indicator (red/green)
  • Mobile: castors with plinth (optionally available), easy moving of the cabinet
  • Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system, extraction of both compartments through one ventilation connection
  • Certificates
    • Fire resistance 90 minutes
    • Type tested in accordance with EN 14470-1
    • EN 16121/16122 conformity
    • CE compliant

Tekniske data

Certifications/Compliance Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant
Configuration 1 x 10 L drawer (sheet steel powder-coated), 1 x pull-out shelf (panel melamine resin-coated/PP)
Depth (Metric) Exterior 502 mm
Depth (Metric) Interior 381 mm
Dimensions (D x W x H) Interior Left: 381 x 470 x 502 mm, Right: 468 x 440 x 562 mm
Dimensions (D x W x H) 502 x 1102 x 601 mm
Door Style Wing
For Use With (Application) Safe and approved storage of aggressive and flammable hazardous substances in working areas
Height (Metric) Exterior 601 mm
Height (Metric) Interior 502 mm
Model K90.060.110.050.UB.ST
No. of Doors 1
No. of Shelves 1
Shelf Type Pull-out
Surface Coating Powder-coated plain
Weight (Metric) 152.50 kg
Width (Metric) Exterior 1102 mm
Width (Metric) Interior 470 mm
Color Light Gray
Depth (English) Exterior 19.76 in.
Height (English) Exterior 23.66 in.
Height (Metric) Opening 502 mm
Material Steel
Product Type Combi Safety Storage Cabinet
Width (English) Exterior 43.39 in.
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.