missing translation for 'onlineSavingsMsg'
Learn More

Kaempferol Tri-O-methoxymethyl Ether, TRC

Product Code. 30395293
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395293 25 mg 25mg
30407879 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395293 Supplier Toronto Research Chemicals Supplier No. TRCK10003025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Kaempferol Tri-O-methoxymethyl Ether
CAS 143724-66-7
Molecular Formula C21H22O9
Quantity 25 mg
Synonym 5-Hydroxy-3,7-bis(methoxymethoxy)-2-[4-(methoxymethoxy)phenyl]-4H-1-benzopyran-4-one
InChI Formula InChI=1S/C21H22O9/c1-24-10-27-14-6-4-13(5-7-14)20-21(29-12-26-3)19(23)18-16(22)8-15(28-11-25-2)9-17(18)30-20/h4-9,22H,10-12H2,1-3H3
SMILES COCOC1=C(Oc2cc(OCOC)cc(O)c2C1=O)c3ccc(OCOC)cc3
IUPAC Name 5-hydroxy-3,7-bis(methoxymethoxy)-2-[4-(methoxymethoxy)phenyl]chromen-4-one
Molecular Weight (g/mol) 418.39
Formula Weight 418.1264
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.